KEY250700003 Key Organics / BIONET

6-(2-methyl-1,3-dioxolan-2-yl)pyridine-2-carboxylic acid

  • MDL number MFCD14584843
  • Cas Number : 122637-38-1
  • Molecular Weight : 209.201

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 113 - 115
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/102236

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/MD-0730_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart