KEY250700019 Key Organics / BIONET

N-[4-bromo-3-(trifluoromethyl)phenyl]-2,2,2-trifluoroacetamide

  • MDL number MFCD00607205
  • Cas Number : 156425-50-2
  • Molecular Weight : 336.031

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 80 - 82
Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94624

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AE-0032_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart