KEY250700023 Key Organics / BIONET

1H,2H,6H,7H,8H-indeno[5,4-b]furan-8-one

  • MDL number MFCD09955085
  • Cas Number : 196597-78-1
  • Molecular Weight : 174.199

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 133 - 135
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/102200

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/MD-0210_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart