KEY250700029 Key Organics / BIONET

1-thia-4-azaspiro[4.5]decan-3-one

  • MDL number MFCD00101403
  • Cas Number : 4580-63-6
  • Molecular Weight : 171.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 166 - 168
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94670

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AE-0714_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart