KEY250700030 Key Organics / BIONET

2-hydroxyphenyl benzoate

  • MDL number MFCD00089689
  • Cas Number : 5876-92-6
  • Molecular Weight : 214.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 114 - 116
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94618

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AE-0005_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart