KEY250700032 Key Organics / BIONET

3-nitrobenzene-1,2-diol

  • MDL number MFCD00463758
  • Cas Number : 6665-98-1
  • Molecular Weight : 155.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 85 - 86
Iupac Name 3-nitro-1,2-benzenediol
Inchi Code 1S/C6H5NO4/c8-5-3-1-2-4(6(5)9)7(10)11/h1-3,8-9H
Inchi Key YHKWFDPEASWKFQ-UHFFFAOYSA-N

Description

Melting Point 85 - 86
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94620

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/AE-0007_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart