KEY267019567 Key Organics / BIONET

2-bromo-3-[(tert-butyldimethylsilyl)oxy]-6-hydroxybenzaldehyde

  • MDL number MFCD16140269
  • Cas Number : 347840-64-6
  • Molecular Weight : 331.281

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 44 - 46
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94627

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AE-0054_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart