KEY267019568 Key Organics / BIONET

3-amino-5-chloro-2-hydroxyphenyl benzoate

  • MDL number MFCD16140259
  • Cas Number : 1221791-82-7
  • Molecular Weight : 263.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 169 - 170
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94638

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AE-0094_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart