KEY267019590 Key Organics / BIONET

2-amino-6-bromo-4-methoxyphenol

  • MDL number MFCD03695460
  • Cas Number : 206872-01-7
  • Molecular Weight : 218.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 100 - 102
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94715

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AE-0773_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart