KEY267019591 Key Organics / BIONET

(3R,4R)-1-benzyl-4-(trifluoromethyl)pyrrolidine-3-carboxylic acid

  • MDL number MFCD13193733
  • Cas Number : 184844-96-0
  • Molecular Weight : 273.255

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 92 Dec
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94659

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AE-0247_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart