KEY267019595 Key Organics / BIONET

[(1R,5S,6R)-3-benzyl-3-azabicyclo[3.1.0]hexan-6-yl]methanol

  • MDL number MFCD16140332
  • Cas Number : 134575-07-8
  • Molecular Weight : 203.285

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point Oil
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/95735

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/CE-0223_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart