KEY267019642 Key Organics / BIONET

(1-methyl- 1,4,5,6 tetrahyrdo-pyrimidin-2-yl)-methylamine dihydrochloride

  • MDL number MFCD16090014
  • Cas Number : 73706-72-6
  • Molecular Weight : 200.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Store under N2
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/108404

Safety Information

signalword Warning
pcodes P422, P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/SS-3831_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart