KEY267019647 Key Organics / BIONET

1-(4-piperidyl)-1,2,3,4-tetrahydro-2-quinolinone hydrochloride

  • MDL number MFCD16090018
  • Cas Number : 141134-83-0
  • Molecular Weight : 266.77

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/108428

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-3875_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart