KEY267019683 Key Organics / BIONET

BOC-(aminophenyl)acetic acid

  • MDL number MFCD09750482
  • Cas Number : 135807-51-1
  • Molecular Weight : 251.282

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/108401

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-3824_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart