KEY267019692 Key Organics / BIONET

3-hydroxybenzamidine hydrochloride

  • MDL number MFCD09954378
  • Cas Number : 63502-89-6
  • Molecular Weight : 172.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/108411

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-3847_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart