KEY267019699 Key Organics / BIONET

4-Spiro-[3-phthalide]piperidine hydrochloride

  • MDL number MFCD02179149
  • Cas Number : 172733-79-8
  • Molecular Weight : 239.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3H-spiro[isobenzofuran-1,4'-piperidin]-3-one hydrochloride
Inchi Code 1S/C12H13NO2.ClH/c14-11-9-3-1-2-4-10(9)12(15-11)5-7-13-8-6-12;/h1-4,13H,5-8H2;1H
Inchi Key JDUFOCPANLQXPS-UHFFFAOYSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/108420

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/SS-3861_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart