KEY267019712 Key Organics / BIONET

2-(4-{[(dimethylamino)carbonyl]amino}phenyl)acetic acid

  • MDL number MFCD14596096
  • Cas Number : 1221792-49-9
  • Molecular Weight : 222.244

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/108434

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-3888_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart