KEY267019715 Key Organics / BIONET

N-(methylsulfonyl)-DL-phenylalanine

  • MDL number MFCD06434623
  • Cas Number : 29268-15-3
  • Molecular Weight : 243.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/108439

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-3895_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart