KEY267019718 Key Organics / BIONET

3-(2-aminoethyl)-7-methyl-imidazo[1,2-A]pyridine

  • MDL number MFCD09971796
  • Cas Number : 171346-86-4
  • Molecular Weight : 175.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Store under N2
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/108443

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404,P422,P422
hcodes H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/SS-3901_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart