KEY267019729 Key Organics / BIONET

4-(acetamidophenylsulfonyl)acetonitrile

  • MDL number MFCD00019833
  • Cas Number : 90915-78-9
  • Molecular Weight : 238.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/108454

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/SS-3915_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart