KEY267019749 Key Organics / BIONET

1H-1,2,3 Triazole-1- acetic acid Hydrochloride

  • MDL number MFCD09029581
  • Cas Number : 1187582-48-4
  • Molecular Weight : 163.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/108489

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-3969_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart