KEY267019756 Key Organics / BIONET

3-(3-(dimethylamino)-3-oxopropyl)benzoic acid

  • MDL number MFCD16090033
  • Cas Number : 940099-04-7
  • Molecular Weight : 221.256

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/108498

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-3990_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart