KEY267019761 Key Organics / BIONET

N-ethyl-3-(hydroxymethyl)piperidine-1-carboxamide

  • MDL number MFCD12079118
  • Cas Number : 940077-81-6
  • Molecular Weight : 186.255

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/108503

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-4001_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart