KEY267019770 Key Organics / BIONET

1-ethyl-3-(5-hydroxypentyl)urea

  • MDL number MFCD16090038
  • Cas Number : 939901-50-5
  • Molecular Weight : 174.244

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/108512

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-4017_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart