KEY267019865 Key Organics / BIONET

1-thia-4-azaspiro[4.4]nonane hydrochloride

  • MDL number MFCD16140324
  • Cas Number : 1221792-36-4
  • Molecular Weight : 179.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 140 - 142
Purity 95
Storage temp. Store under N2, Hygroscopic
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94674

Safety Information

signalword Warning
pcodes P232,P422, P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AE-0719_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart