KEY267019868 Key Organics / BIONET

8-amino-6,7-diazaspiro[3.4]oct-7-en-5-one

  • MDL number MFCD16140329
  • Cas Number : 1221791-84-9
  • Molecular Weight : 139.158

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 86 - 88
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94702

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AE-0756_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart