KEY267019870 Key Organics / BIONET

methyl 1-oxo-3,4-dihydro-2H-1lambda4,4-benzothiazine-6-carboxylate

  • MDL number MFCD16140306
  • Cas Number : 1221792-60-4
  • Molecular Weight : 225.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 158 - 162
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94705

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AE-0760_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart