KEY267019872 Key Organics / BIONET

5-bromo-3-cyano-4,6-dimethylpyridine-2-carboxylic acid

  • MDL number MFCD16140267
  • Cas Number : 1221792-64-8
  • Molecular Weight : 255.071

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 187 Dec
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94710

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AE-0766_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart