KEY267019877 Key Organics / BIONET

methyl 5-bromo-2-oxo-2,3-dihydro-1,3-benzoxazole-7-carboxylate

  • MDL number MFCD16140296
  • Cas Number : 1221792-66-0
  • Molecular Weight : 272.054

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 240 - 242
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/95070

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BE-0036_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart