KEY267019924 Key Organics / BIONET

2-(azetidin-3-yl)pyridine dihydrochloride

  • MDL number MFCD16140331
  • Cas Number : 206446-45-9
  • Molecular Weight : 207.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 174 - 175
Purity 95
Storage temp. Store under N2, Hygroscopic
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/95733

Safety Information

signalword Warning
pcodes P232,P422, P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/CE-0219_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart