KEY267019946 Key Organics / BIONET

methyl 5-methyl-2-oxo-2,3-dihydro-1,3-benzoxazole-7-carboxylate

  • MDL number MFCD16140305
  • Cas Number : 1221792-45-5
  • Molecular Weight : 207.185

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 234 - 236
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/106752

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/ND-0070_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart