KEY267019947 Key Organics / BIONET

N-(9-fluorenylmethoxycarbonyl)-1,2,3,4-tetrahydro-1-naphthylamine-4-carboxylic acid

  • MDL number MFCD16140289
  • Cas Number : 1221792-81-9
  • Molecular Weight : 413.473

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form White to Off-white Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/108407

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/SS-3840_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart