KEY267019952 Key Organics / BIONET

3-(4-bromophenyl)-4-methyl-4,5-dihydro-1H-1,2,4-triazole-5-thione

  • MDL number MFCD03995808
  • Cas Number : 494194-50-2
  • Molecular Weight : 270.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 246 - 248
Iupac Name 5-(4-bromophenyl)-4-methyl-4H-1,2,4-triazol-3-yl hydrosulfide
Inchi Code 1S/C9H8BrN3S/c1-13-8(11-12-9(13)14)6-2-4-7(10)5-3-6/h2-5H,1H3,(H,12,14)
Inchi Key FHXAECCDIGEICD-UHFFFAOYSA-N

Description

Melting Point 246 - 248
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/95707

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/CE-0050_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart