KEY267019954 Key Organics / BIONET

1H,2H,3H,4H,5H,6H,7H-cyclopenta[d]pyrimidine-2,4-dione

  • MDL number MFCD00466420
  • Cas Number : 5466-00-2
  • Molecular Weight : 152.153

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 328 - 330
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/95127

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BE-0739_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart