KEY267019959 Key Organics / BIONET

2-(2-phenylethyl)piperazine

  • MDL number MFCD07373378
  • Cas Number : 91907-37-8
  • Molecular Weight : 190.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 101 - 102
Iupac Name 2-(2-phenylethyl)piperazine
Inchi Code 1S/C12H18N2/c1-2-4-11(5-3-1)6-7-12-10-13-8-9-14-12/h1-5,12-14H,6-10H2
Inchi Key FRTXUSHWBSBIAT-UHFFFAOYSA-N

Description

Melting Point 101 - 102
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/95730

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/CE-0213_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart