KEY267019961 Key Organics / BIONET

1,3-thiazolidin-2-one

  • MDL number MFCD01332388
  • Cas Number : 2682-49-7
  • Molecular Weight : 103.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 42 - 44
Iupac Name 1,3-thiazolidin-2-one
Inchi Code 1S/C3H5NOS/c5-3-4-1-2-6-3/h1-2H2,(H,4,5)
Inchi Key SLYRGJDSFOCAAI-UHFFFAOYSA-N

Description

Melting Point 42 - 44
Purity 95%
Physical Form Low Melting Solid,Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/95129

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/BE-0742_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart