KEY267019962 Key Organics / BIONET

tert-butyl 4-(2-ethoxy-2-oxoethylidene)piperidine-1-carboxylate

  • MDL number MFCD12912655
  • Cas Number : 135716-08-4
  • Molecular Weight : 269.341

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 88 - 89
Purity 95
Storage temp. RT
Physical Form White to Off-white Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/95737

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/CE-0229_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart