KEY267019976 Key Organics / BIONET

tert-butyl 3-(hydrazinecarbonyl)piperidine-1-carboxylate

  • MDL number MFCD06795929
  • Cas Number : 625470-88-4
  • Molecular Weight : 243.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp Oil
Iupac Name tert-butyl 3-(hydrazinocarbonyl)-1-piperidinecarboxylate
Inchi Code 1S/C11H21N3O3/c1-11(2,3)17-10(16)14-6-4-5-8(7-14)9(15)13-12/h8H,4-7,12H2,1-3H3,(H,13,15)
Inchi Key DABYYYLRDBQJTK-UHFFFAOYSA-N

Description

Melting Point Oil
Purity 97%
Physical Form Low Melting Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/95139

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/BE-0754_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart