KEY267019977 Key Organics / BIONET

5-nitro-2-[(propan-2-yl)amino]benzoic acid

  • MDL number MFCD11131983
  • Cas Number : 1019387-83-7
  • Molecular Weight : 224.216

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 229 - 230
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94700

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AE-0753_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart