KEY267019989 Key Organics / BIONET

methyl 4-amino-5-iodo-2-methoxybenzoate

  • MDL number MFCD11112895
  • Cas Number : 137832-56-5
  • Molecular Weight : 307.087

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 160 - 164
Purity 95
Storage temp. RT
Physical Form Off-white to Dark Orange Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/95148

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/BE-0768_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart