KEY267019991 Key Organics / BIONET

6-chloro-N-methoxy-N-methylpyridine-3-carboxamide

  • MDL number MFCD09991647
  • Cas Number : 149281-42-5
  • Molecular Weight : 200.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 106 - 110 @ 0.5 mm Hg
Iupac Name 6-chloro-N-methoxy-N-methylnicotinamide
Inchi Code 1S/C8H9ClN2O2/c1-11(13-2)8(12)6-3-4-7(9)10-5-6/h3-5H,1-2H3
Inchi Key IJUKASNMWCZHHA-UHFFFAOYSA-N

Description

Melting Point 106 - 110 @ 0.5 mm Hg
Purity 95%
Physical Form Liquid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94709

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/AE-0765_SDS.pdf
hcodes H302,H312,H315,H319,H332,H335
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart