KEY267019999 Key Organics / BIONET

2-N-Fmoc-amino-4,5-dimethoxybenzoic acid

  • MDL number MFCD02682187
  • Cas Number : 855005-12-8
  • Molecular Weight : 419.433

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/108413

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-3850_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart