KEY267020010 Key Organics / BIONET

FMOC-DL-2-pyridylalanine

  • MDL number MFCD02094443
  • Cas Number : 185379-40-2
  • Molecular Weight : 388.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/108464

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/SS-3927_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart