KEY267020013 Key Organics / BIONET

FMOC-DL-3-benzothienylalanine

  • MDL number MFCD03412494
  • Cas Number : 177966-60-8
  • Molecular Weight : 443.52

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/108467

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/SS-3931_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart