KEY267020015 Key Organics / BIONET

ethyl (2E)-3-(pyridin-2-yl)prop-2-enoate

  • MDL number MFCD16140351
  • Cas Number : 70526-11-3
  • Molecular Weight : 177.203

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 75 @ 0.02 mBar
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/95087

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BE-0204_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart