KEY293014780 Key Organics / BIONET

tert-butyl 1,9-diazaspiro[5.5]undecane-9-carboxylate

  • MDL number MFCD09998745
  • Cas Number : 1031927-14-6
  • Molecular Weight : 254.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name tert-butyl 1,9-diazaspiro[5.5]undecane-9-carboxylate
Inchi Code 1S/C14H26N2O2/c1-13(2,3)18-12(17)16-10-7-14(8-11-16)6-4-5-9-15-14/h15H,4-11H2,1-3H3
Inchi Key FYXOOJBRPNJJRP-UHFFFAOYSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/96157

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/DE-0047_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart