KEY293014782 Key Organics / BIONET

methyl 2-chloro-5-nitropyridine-3-carboxylate

  • MDL number MFCD07368888
  • Cas Number : 190271-88-6
  • Molecular Weight : 216.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 62 - 64
Iupac Name methyl 2-chloro-5-nitronicotinate
Inchi Code 1S/C7H5ClN2O4/c1-14-7(11)5-2-4(10(12)13)3-9-6(5)8/h2-3H,1H3
Inchi Key IPSMDDVNXJKYRH-UHFFFAOYSA-N

Description

Melting Point 62 - 64
Purity 97%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/96473

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/EE-0704_SDS.pdf
hcodes H302,H312,H315,H319,H332,H335
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart