KEY293014902 Key Organics / BIONET

2-hydroxy-3-methoxy-3,3-diphenylpropanoic acid

  • MDL number MFCD13152279
  • Cas Number : 178306-51-9
  • Molecular Weight : 272.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point Decomp > 95
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/95742

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/CE-0241_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart