KEY293014905 Key Organics / BIONET

1-(2-cyanophenyl)-3-(prop-2-en-1-yl)urea

  • MDL number MFCD16622763
  • Cas Number : 122372-27-4
  • Molecular Weight : 201.229

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 173 - 175
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/1063

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/10H-440S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart