KEY293014909 Key Organics / BIONET

methyl 2,4-dioxo-1,2,3,4-tetrahydroquinazoline-5-carboxylate

  • MDL number MFCD13250068
  • Cas Number : 127801-84-7
  • Molecular Weight : 220.184

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 312 - 314
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/96472

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/EE-0703_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart